Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026499.4 | NP_080775.3 | 4 | 688 | ||
GenBank mRNA | AK004831.1 | - | 4 | 669 | ||
AK012605.1 | - | 4 | 436 | |||
AK051608.1 | - | 2 | 841 | |||
AK133872.1 | - | 4 | 669 | |||
AK146293.1 | - | 4 | 666 | |||
AK159519.1 | - | 4 | 666 | |||
AK171709.1 | - | 3 | 1952 | |||
AK179065.1 | - | 3 | 317 | |||
AK208133.1 | - | 1 | 135 | |||
BC012039.1 | - | 4 | 675 | 2648111 |