Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2700062C07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026529.4 | NP_080805.1 | 4 | 497 | ||
GenBank mRNA | AK011228.1 | - | 3 | 584 | ||
AK012467.1 | - | 3 | 599 | |||
AK013030.1 | - | 4 | 405 | |||
AK018391.1 | - | 4 | 501 | |||
AK164298.1 | - | 2 | 2200 | |||
BC029246.1 | - | 4 | 1154 | |||
BC037075.1 | - | 4 | 545 | 5038294 | ||
BC084681.1 | - | 4 | 507 | 30144888 |