Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpnd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026530.5 | NP_080806.4 | 6 | 772 | ||
GenBank mRNA | AK012128.1 | - | 5 | 727 | ||
AK146973.1 | - | 6 | 757 | |||
AK160356.1 | - | 6 | 741 | |||
AK172381.1 | - | 5 | 515 | |||
AK213160.1 | - | 4 | 261 | |||
BC017625.1 | - | 5 | 721 |