Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026539.2 | NP_080815.1 | 22 | 2567 | ||
GenBank mRNA | AK014473.1 | - | 22 | 2569 | ||
AK165656.1 | - | 22 | 2594 | |||
BC020486.1 | - | 7 | 739 | |||
BC052385.1 | - | 22 | 2551 | 5703859 | ||
BC057567.1 | - | 22 | 2580 | 6415898 | ||
BC062966.1 | - | 22 | 2580 | 6415898 |