Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dynll2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168471.1 | NP_001161943.1 | 3 | 1188 | ||
NM_001168472.1 | NP_001161944.1 | 3 | 1361 | |||
NM_026556.4 | NP_080832.1 | 3 | 1394 | |||
GenBank mRNA | AK028319.1 | - | 3 | 1159 | ||
AK049645.1 | - | 3 | 1368 | |||
AK078434.1 | - | 3 | 1367 | |||
AK160114.1 | - | 3 | 1364 | |||
AK169212.1 | - | 3 | 1368 | |||
BC011289.1 | - | 3 | 1181 | 3981362 | ||
BC040822.1 | - | 3 | 1352 | 4459858 | ||
BC058797.1 | - | 3 | 1000 |