Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D730048I06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026593.3 | NP_080869.1 | 2 | 174 | ||
GenBank mRNA | AK020299.1 | - | 2 | 146 | ||
AK020302.1 | - | 2 | 146 | |||
AK020310.1 | - | 2 | 127 | |||
AK021351.1 | - | 2 | 146 | |||
AK033813.1 | - | 2 | 175 | |||
AK136605.1 | - | 164 | ||||
AK136636.1 | - | 2 | 145 | |||
BC027498.1 | - | 2 | 108 | 3465762 |