Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183270.2 | NP_899093.1 | 2 | 709 | ||
GenBank mRNA | AK019895.1 | - | 681 | |||
AK088786.1 | - | 2 | 697 | |||
AK161506.1 | - | 710 | ||||
BC048574.1 | - | 2 | 650 |