Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam135a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026604.4 | NP_080880.4 | 20, 21 | 4735 | ||
GenBank mRNA | AK019549.1 | - | 7, 8 | 2353 | ||
AK034175.1 | - | 7, 8 | 3101 | |||
AK044359.1 | - | 7, 8 | 2899 | |||
AK087116.1 | - | 7, 8 | 923 | |||
AK142968.1 | - | 6, 7 | 621 | |||
AK173166.1 | - | 4, 5 | 5495 | |||
BC031160.1 | - | 7, 8 | 2757 | |||
BC039976.1 | - | 5, 6 | 561 | |||
BC066812.1 | - | 7, 8 | 1678 | |||
BC070446.1 | - | 19, 20 | 4417 | 30362784 | ||
BC083093.1 | - | 7, 8 | 2214 |