Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam135a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026604.4 | NP_080880.4 | 18 | 4378 | ||
GenBank mRNA | AK012197.1 | - | 5 | 1861 | ||
AK019549.1 | - | 5 | 1996 | |||
AK034175.1 | - | 5 | 2744 | |||
AK044359.1 | - | 5 | 2542 | |||
AK087116.1 | - | 5 | 565 | |||
AK142968.1 | - | 4 | 264 | |||
AK173166.1 | - | 2 | 5138 | |||
BC031160.1 | - | 5 | 2400 | |||
BC039976.1 | - | 3 | 204 | |||
BC066812.1 | - | 5 | 1321 | |||
BC070446.1 | - | 17 | 4060 | 30362784 | ||
BC083093.1 | - | 5 | 1857 |