Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024221.3 | NP_077183.1 | 7 | 723 | ||
GenBank mRNA | AK011810.1 | - | 687 | |||
AK153058.1 | - | 7 | 677 | |||
AK166631.1 | - | 7 | 725 | |||
AK167798.1 | - | 7 | 679 | |||
AK218654.1 | - | 2 | 262 | |||
BC002188.1 | - | 5 | 547 | |||
BC019512.1 | - | 7 | 696 | 4210204 | ||
BC094468.1 | - | 7 | 669 | 5011266 |