Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Toporsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026652.2 | NP_080928.1 | 2 | 461 | ||
GenBank mRNA | AK015747.1 | - | 2 | 495 | ||
AK019748.1 | - | 2 | 462 | |||
BC120821.1 | - | 1 | 330 | 40129744 | ||
BC120823.1 | - | 1 | 330 | 40129746 |