Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MEF2C SIRNA Assays ›
Gene Symbol:
MEF2C
Gene Name:
myocyte enhancer factor 2C
Gene Aliases:
C5DELq14.3, DEL5q14.3
Chromosome Location:
Chr.5: 88718241 - 88904105 on Build GRCh38
UniGene ID:
Hs.649965
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s8653
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MEF2C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001131005.2
NP_001124477.1
8
1175
NM_001193347.1
NP_001180276.1
9
1235
NM_001193348.1
NP_001180277.1
5
747
NM_001193349.1
NP_001180278.1
5
747
NM_001193350.1
NP_001180279.1
7
1403
NM_001308002.1
NP_001294931.1
6
891
NM_002397.4
NP_002388.2
7
1406
XM_005248511.2
XP_005248568.1
7
940
XM_006714619.2
XP_006714682.1
8
1262
XM_006714625.3
XP_006714688.1
8
1293
XM_011543396.2
XP_011541698.1
7
991
XM_011543397.2
XP_011541699.1
7
1172
XM_011543400.1
XP_011541702.1
7
1178
XM_011543401.1
XP_011541703.1
7
1178
XM_017009475.1
XP_016864964.1
7
1233
XM_017009476.1
XP_016864965.1
7
1239
XM_017009477.1
XP_016864966.1
7
1178
XM_017009478.1
XP_016864967.1
7
1233
XM_017009479.1
XP_016864968.1
7
1239
XM_017009480.1
XP_016864969.1
8
1262
XM_017009481.1
XP_016864970.1
7
940
XM_017009482.1
XP_016864971.1
7
1239
XM_017009483.1
XP_016864972.1
7
1233
GenBank mRNA
AK307883.1
-
6
748
AK312472.1
-
7
1111
AL833268.1
-
9
1183
AL833274.1
-
8
1124
BC026341.1
-
7
1178
FM163484.2
-
5
608
FM180475.1
-
5
608
L08895.1
-
7
1153
S57212.1
-
1412
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
blood vessel development
osteoblast differentiation
neuron migration
B cell homeostasis
heart looping
endochondral ossification
blood vessel remodeling
chondrocyte differentiation
germinal center formation
regulation of germinal center formation
primary heart field specification
secondary heart field specification
outflow tract morphogenesis
sinoatrial valve morphogenesis
cardiac ventricle formation
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
apoptotic process
humoral immune response
nervous system development
heart development
muscle organ development
skeletal muscle tissue development
muscle cell fate determination
learning or memory
response to virus
positive regulation of gene expression
negative regulation of gene expression
positive regulation of alkaline phosphatase activity
neural crest cell differentiation
cardiac muscle hypertrophy in response to stress
myotube differentiation
neuron differentiation
platelet formation
monocyte differentiation
negative regulation of ossification
melanocyte differentiation
positive regulation of bone mineralization
positive regulation of B cell proliferation
cellular response to drug
skeletal muscle cell differentiation
cellular response to trichostatin A
B cell proliferation
regulation of neuron apoptotic process
negative regulation of neuron apoptotic process
regulation of megakaryocyte differentiation
positive regulation of myoblast differentiation
positive regulation of neuron differentiation
positive regulation of osteoblast differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of neurotransmitter secretion
regulation of synaptic plasticity
positive regulation of skeletal muscle tissue development
neuron development
cell morphogenesis involved in neuron differentiation
embryonic viscerocranium morphogenesis
negative regulation of epithelial cell proliferation
B cell receptor signaling pathway
smooth muscle cell differentiation
positive regulation of muscle cell differentiation
regulation of synapse assembly
regulation of synaptic transmission, glutamatergic
ventricular cardiac muscle cell differentiation
palate development
regulation of synaptic activity
positive regulation of cardiac muscle cell proliferation
excitatory postsynaptic potential
regulation of sarcomere organization
cartilage morphogenesis
regulation of dendritic spine development
renal tubule morphogenesis
cellular response to lipopolysaccharide
cellular response to calcium ion
cellular response to parathyroid hormone stimulus
cellular response to fluid shear stress
cellular response to transforming growth factor beta stimulus
positive regulation of cell proliferation in bone marrow
glomerulus morphogenesis
nephron tubule epithelial cell differentiation
positive regulation of protein homodimerization activity
positive regulation of macrophage apoptotic process
regulation of N-methyl-D-aspartate selective glutamate receptor activity
regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
positive regulation of cardiac muscle cell differentiation
positive regulation of behavioral fear response
epithelial cell proliferation involved in renal tubule morphogenesis
positive regulation of skeletal muscle cell differentiation
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II core promoter sequence-specific
core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
AT DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
activating transcription factor binding
miRNA binding
histone deacetylase binding
transcription regulatory region DNA binding
protein heterodimerization activity
HMG box domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-k9xzd:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0