Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aph1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026674.3 | NP_080950.1 | 6 | 1271 | ||
GenBank mRNA | BC050923.1 | - | 6 | 1271 | 6410104 | |
BC063254.1 | - | 6 | 1271 | 6410104 |