Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163718.1 | NP_001157190.1 | 6 | 946 | ||
NM_026676.3 | NP_080952.2 | 6 | 967 | |||
GenBank mRNA | AK002309.1 | - | 6 | 939 | ||
AK003675.1 | - | 3 | 1715 | |||
AK158743.1 | - | 6 | 968 | |||
AK169072.1 | - | 5 | 1302 | |||
AK170074.1 | - | 6 | 936 | |||
BC006899.1 | - | 3 | 312 | |||
BC027751.1 | - | 6 | 1037 | 5011894 | ||
BC038884.1 | - | 3 | 659 | 1516275 | ||
BC089019.1 | - | 6 | 916 | 30551363 |