Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ABL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005157.5 | NP_005148.2 | 2 | 434 | ||
NM_007313.2 | NP_009297.2 | 2 | 738 | |||
GenBank mRNA | AB209456.1 | - | 2 | 444 | ||
AB209642.1 | - | 2 | 667 | |||
BC117451.1 | - | 2 | 364 | 40126002 | ||
M14752.1 | - | 2 | 606 | |||
M30833.1 | - | 2 | 389 | |||
X16416.1 | - | 2 | 389 |