Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291235.1 | NP_001278164.1 | 4 | 283 | ||
NM_001291236.1 | NP_001278165.1 | 4 | 759 | |||
NM_001291237.1 | NP_001278166.1 | 5 | 467 | |||
NM_026708.2 | NP_080984.1 | 4 | 594 | |||
GenBank mRNA | AK002265.1 | - | 4 | 542 | ||
AK046577.1 | - | 5 | 2474 | |||
AK161721.1 | - | 494 | ||||
AK165432.1 | - | 706 | ||||
BC005702.1 | - | 4 | 486 | 3598444 | ||
BC039182.1 | - | 4 | 729 | 3500908 | ||
BG083251.2 | - | 4 | 341 |