Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6pc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175935.3 | NP_787949.2 | 4 | 717 | ||
GenBank mRNA | AK002966.1 | - | 5 | 580 | ||
AK165395.1 | - | 4 | 571 | |||
AY186239.1 | - | 4 | 563 | |||
BC069959.1 | - | 4 | 717 | 4990136 |