Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsmce2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164604.1 | NP_001158076.1 | 3, 4 | 434 | ||
NM_026746.3 | NP_081022.2 | 3, 4 | 434 | |||
GenBank mRNA | AK003685.1 | - | 3, 4 | 386 | ||
AK051277.1 | - | 3, 4 | 386 | |||
AK211545.1 | - | 315 | ||||
AK212165.1 | - | 315 | ||||
BC009125.1 | - | 3, 4 | 345 | 3155917 | ||
DV057668.1 | - | 3, 4 | 454 |