Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190297.2 | NP_001177226.1 | 3 | 513 | ||
NM_001276443.1 | NP_001263372.1 | 3 | 513 | |||
NM_001276444.1 | NP_001263373.1 | 3 | 513 | |||
GenBank mRNA | AK089436.1 | - | 3 | 431 | ||
AK090081.1 | - | 4 | 517 | |||
BC115698.1 | - | 3 | 409 | 40060105 | ||
HM161763.2 | - | 3 | 490 | |||
HM161764.2 | - | 3 | 490 | |||
HM161765.2 | - | 3 | 490 | |||
HM161766.2 | - | 3 | 490 | |||
KF358283.1 | - | 3 | 490 | |||
KF358284.1 | - | 3 | 490 |