Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2310036O22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026760.2 | NP_081036.2 | 2 | 430 | ||
GenBank mRNA | AK003299.1 | - | 2 | 395 | ||
AK009653.1 | - | 2 | 397 | |||
BC023284.1 | - | 2 | 440 | 3484935 | ||
BC029622.1 | - | 2 | 430 | 4207296 |