Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024226.4 | NP_077188.1 | 4 | 682 | ||
NM_194051.3 | NP_918940.1 | 6 | 2969 | |||
NM_194052.3 | NP_918941.1 | 4 | 1158 | |||
NM_194053.3 | NP_918942.1 | 5 | 1215 | |||
NM_194054.3 | NP_918943.1 | 6 | 3576 | |||
GenBank mRNA | AB073672.1 | - | 4 | 1534 | ||
AF326337.1 | - | 4 | 462 | |||
AK212390.1 | - | 4 | 385 | |||
AK218123.1 | - | 4 | 387 | |||
AK220511.1 | - | 6 | 3536 | |||
AY102280.1 | - | 6 | 2969 | |||
AY102281.1 | - | 4 | 1115 | |||
AY102282.1 | - | 5 | 1172 | |||
AY102283.1 | - | 4 | 675 | |||
AY102284.1 | - | 6 | 3533 | |||
AY114152.1 | - | 6 | 3534 | |||
AY114153.1 | - | 5 | 989 | |||
BC032192.1 | - | 4 | 1828 | 5323152 | ||
BC032272.1 | - | 4 | 2682 | |||
BC056373.1 | - | 6 | 3404 | |||
BU848611.1 | - | 4 | 521 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK082222.1 | - | 1684 | Chr. - - [-] on Build |