Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026814.3 | NP_081090.1 | 2 | 401 | ||
GenBank mRNA | AK004064.1 | - | 2 | 402 | ||
BC115957.1 | - | 2 | 382 | 40055543 |