Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190437.1 | NP_001177366.1 | 5 | 574 | ||
NM_145215.2 | NP_660250.1 | 5 | 1051 | |||
GenBank mRNA | AF412033.1 | - | 5 | 1051 | ||
AK004244.1 | - | 5 | 585 | |||
BC016241.1 | - | 3 | 625 | |||
BC038052.1 | - | 4 | 453 | |||
BC056610.1 | - | 5 | 713 | |||
BC056624.1 | - | 4 | 1036 | |||
BC069865.1 | - | 2 | 1038 | |||
BC069866.1 | - | 5 | 673 | 6478980 |