Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Abhd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190437.1 | NP_001177366.1 | 2 | 79 | ||
NM_145215.2 | NP_660250.1 | 2 | 556 | |||
GenBank mRNA | AF412033.1 | - | 2 | 556 | ||
AK004244.1 | - | 2 | 91 | |||
BC056610.1 | - | 2 | 249 | |||
BC056624.1 | - | 1 | 541 | |||
BC069865.1 | - | 1 | 291 | |||
BC069866.1 | - | 2 | 178 | 6478980 |