Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK037622.1 | - | 7 | 1110 | ||
BC172146.1 | - | 7 | 858 | |||
DQ924536.1 | - | 7 | 1169 |