Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293763.1 | NP_001280692.1 | 3 | 1221 | ||
NM_001293766.1 | NP_001280695.1 | 2 | 1088 | |||
NM_001293767.1 | NP_001280696.1 | 3 | 1080 | |||
NM_001293768.1 | NP_001280697.1 | 3 | 1497 | |||
NM_026869.3 | NP_081145.1 | 3 | 1163 | |||
GenBank mRNA | AK132759.1 | - | 3 | 1164 | ||
AK186308.1 | - | 1 | 98 | |||
AK202034.1 | - | 11 | ||||
AK204574.1 | - | 11 | ||||
AK204606.1 | - | 11 | ||||
AK206812.1 | - | 11 | ||||
AK219319.1 | - | 11 | ||||
AY486147.1 | - | 3 | 1450 | |||
BC025531.1 | - | 1 | 914 | 5250918 | ||
BC025640.1 | - | 3 | 1153 | 5322622 | ||
BC030445.1 | - | 1 | 598 |