Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Chmp2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026879.2 | NP_081155.1 | 3 | 389 | ||
GenBank mRNA | AK084111.1 | - | 3 | 391 | ||
AK180291.1 | - | 3 | 208 | |||
AK197280.1 | - | 3 | 208 | |||
AK201812.1 | - | 3 | 206 | |||
AK217905.1 | - | 3 | 209 | |||
BC042626.1 | - | 2 | 145 | |||
BC055809.1 | - | 3 | 418 | 6414077 |