Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013256.2 | NP_001013274.2 | 17 | 3053 | ||
NM_001143790.1 | NP_001137262.1 | 17 | 3050 | |||
NM_001281464.1 | NP_001268393.1 | 17 | 2921 | |||
NM_001281465.1 | NP_001268394.1 | 17 | 3037 | |||
GenBank mRNA | AK038651.1 | - | 17 | 2837 | ||
AK044055.1 | - | 17 | 2832 | |||
AK145919.1 | - | 17 | 2868 | |||
AK165906.1 | - | 17 | 2853 | |||
AK176964.1 | - | 1 | 77 | |||
AK186509.1 | - | 1 | 101 | |||
AK198189.1 | - | 3 | 326 | |||
AK209872.1 | - | 1 | 101 | |||
AK213640.1 | - | 1 | 101 | |||
AK214175.1 | - | 2 | 173 | |||
AK217143.1 | - | 1 | 101 | |||
BC014687.1 | - | 8 | 1221 | |||
BC060629.1 | - | 13 | 2103 | 6847619 | ||
BC066078.1 | - | 17 | 2864 | |||
BC072625.1 | - | 17 | 2878 | |||
BC147495.1 | - | 17 | 2851 | 9056325 | ||
BC147497.1 | - | 17 | 2851 | 9056354 |