Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159714.1 | NP_001153186.1 | 12, 13 | 1342 | ||
NM_027328.4 | NP_081604.3 | 12, 13 | 1360 | |||
GenBank mRNA | AK005294.1 | - | 12, 13 | 1327 | ||
AK033190.1 | - | 12, 13 | 1363 | |||
AK033283.1 | - | 12, 13 | 1325 | |||
AK051260.1 | - | 12, 13 | 1320 | |||
AY040823.1 | - | 12, 13 | 1336 | |||
BC018376.1 | - | 12, 13 | 1326 | 3673893 | ||
BC057877.1 | - | 11, 12 | 1303 | 5347609 | ||
BC061461.1 | - | 12, 13 | 1286 | 5717606 | ||
CT010189.1 | - | 11, 12 | 1280 |