Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026912.1 | NP_081188.1 | 2, 3 | 199 | ||
GenBank mRNA | BC016091.1 | - | 2, 3 | 199 | 4503873 |