Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pycr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133705.2 | NP_598466.1 | 6, 7 | 980 | ||
GenBank mRNA | AK007535.1 | - | 6, 7 | 961 | ||
AK150756.1 | - | 4, 5 | 2100 | |||
AK152302.1 | - | 4, 5 | 2100 | |||
AK165618.1 | - | 6, 7 | 955 | |||
AK167622.1 | - | 6, 7 | 936 | |||
AK179626.1 | - | 1, 2 | 21 | |||
AK182076.1 | - | 1, 2 | 21 | |||
AK186434.1 | - | 1, 2 | 21 | |||
AK210838.1 | - | 1, 2 | 21 | |||
BC006882.1 | - | 6, 7 | 956 | 2647612 |