Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fuom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286217.1 | NP_001273146.1 | 6 | 539 | ||
NM_001286218.1 | NP_001273147.1 | 5 | 552 | |||
NM_026928.3 | NP_081204.2 | 6 | 623 | |||
GenBank mRNA | AK007494.1 | - | 6 | 587 | ||
AK044731.1 | - | 6 | 664 | |||
BC028662.1 | - | 6 | 584 | 1332340 | ||
CK618091.1 | - | 6 | 474 |