Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178616.3 | NP_848731.2 | 6 | 518 | ||
GenBank mRNA | AK007547.1 | - | 6 | 487 | ||
AK012951.1 | - | 6 | 527 | |||
AK029365.1 | - | 2 | 89 | |||
AK051738.1 | - | 6 | 508 | |||
AK076002.1 | - | 6 | 506 | |||
AK089942.1 | - | 6 | 483 | |||
AK136049.1 | - | 6 | 481 | |||
AK140773.1 | - | 503 | ||||
AK167559.1 | - | 6 | 489 | |||
AK168289.1 | - | 6 | 510 | |||
AK192313.1 | - | 4 | 201 | |||
BC030432.1 | - | 3 | 172 | |||
BC037590.1 | - | 6 | 467 | |||
BC055457.1 | - | 6 | 450 | |||
BC090664.1 | - | 6 | 470 | 30677672 | ||
BC090980.1 | - | 6 | 485 | 30283128 | ||
BC100595.1 | - | 6 | 480 | 6767671 | ||
BC119136.1 | - | 6 | 489 | 8733885 | ||
BC119138.1 | - | 6 | 489 | 8733887 |