Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alkbh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026944.1 | NP_081220.1 | 6, 7 | 496 | ||
GenBank mRNA | AK007557.1 | - | 16 | |||
AK145586.1 | - | 6, 7 | 882 | |||
BC018196.1 | - | 6, 7 | 496 | 3993214 |