Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cd209b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037800.3 | NP_001032889.3 | 1, 2 | 106 | ||
NM_001287211.1 | NP_001274140.1 | 1, 2 | 106 | |||
NM_026972.5 | NP_081248.4 | 1, 2 | 106 | |||
GenBank mRNA | AF373409.1 | - | 1, 2 | 84 | ||
AF374471.1 | - | 1, 2 | 63 | |||
AF422108.1 | - | 1, 2 | 79 | |||
AF424790.1 | - | 1, 2 | 78 | |||
AF424791.1 | - | 1, 2 | 78 | |||
AF424792.1 | - | 1, 2 | 78 | |||
AF424793.1 | - | 1, 2 | 78 | |||
AF424794.1 | - | 1, 2 | 98 | |||
AF424795.1 | - | 1, 2 | 95 | |||
AF424796.1 | - | 1, 2 | 89 | |||
AF424797.1 | - | 1, 2 | 78 | |||
AF424798.1 | - | 1, 2 | 98 | |||
AF424799.1 | - | 1, 2 | 78 | |||
AF424803.1 | - | 1, 2 | 114 | |||
AK007656.1 | - | 1 | 22 | |||
AK138999.1 | - | 1, 2 | 80 |