Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd209b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037800.3 | NP_001032889.3 | 9 | 879 | ||
NM_001287211.1 | NP_001274140.1 | 10 | 942 | |||
NM_026972.5 | NP_081248.4 | 10 | 969 | |||
GenBank mRNA | AF373409.1 | - | 10 | 947 | ||
AF374471.1 | - | 9 | 836 | |||
AF422108.1 | - | 10 | 942 | |||
AF424791.1 | - | 10 | 941 | |||
AF424792.1 | - | 10 | 941 | |||
AF424793.1 | - | 10 | 941 | |||
AF424794.1 | - | 10 | 961 | |||
AF424795.1 | - | 10 | 958 | |||
AF424796.1 | - | 10 | 952 | |||
AF424797.1 | - | 9 | 851 | |||
AF424798.1 | - | 9 | 871 | |||
AF424799.1 | - | 10 | 914 | |||
AF424800.1 | - | 4 | 372 | |||
AF424801.1 | - | 4 | 372 | |||
AF424802.1 | - | 4 | 372 | |||
AF424803.1 | - | 9 | 825 | |||
AK007656.1 | - | 9 | 885 | |||
AK138999.1 | - | 10 | 943 |