Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028839.4 | NP_083115.3 | 2, 3 | 347 | ||
GenBank mRNA | AK007740.1 | - | 2, 3 | 355 | ||
AK009341.1 | - | 2, 3 | 326 | |||
AK076341.1 | - | 2, 3 | 326 | |||
AK149088.1 | - | 2, 3 | 320 | |||
AK155257.1 | - | 2, 3 | 312 | |||
CJ180452.1 | - | 2, 3 | 351 |