Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026981.2 | NP_081257.1 | 4 | 694 | ||
GenBank mRNA | AK007678.1 | - | 3 | 661 | ||
AK149746.1 | - | 3 | 4244 | |||
BC020149.1 | - | 4 | 641 | 3993232 |