Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038998.2 | NP_001034087.1 | 2 | 339 | ||
NM_024462.2 | NP_077782.1 | 2 | 265 | |||
GenBank mRNA | AI048353.1 | - | 2 | 165 | ||
AK010418.1 | - | 2 | 195 | |||
AK079783.1 | - | 2 | 124 | |||
BC002274.1 | - | 2 | 178 | 3496493 | ||
BF012097.1 | - | 2 | 269 | |||
BQ044357.1 | - | 1 | 31 | |||
CF105092.1 | - | 2 | 325 |