Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027002.3 | NP_081278.1 | 1 | 77 | ||
NM_027101.2 | NP_081377.1 | 1 | 60 | |||
GenBank mRNA | AK009093.1 | - | 1 | 81 | ||
AK011600.1 | - | 1 | 79 | |||
BC004810.1 | - | 1 | 60 | 3588284 |