Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027009.2 | NP_081285.1 | 9 | 1085 | ||
GenBank mRNA | AK145126.1 | - | 9 | 1112 | ||
AK165891.1 | - | 6 | 2596 | |||
AK167047.1 | - | 9 | 1086 | |||
BC026795.1 | - | 9 | 1056 | 4015134 |