Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700001J03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008547.1 | NP_001008547.1 | 2 | 228 | ||
GenBank mRNA | AK005597.1 | - | 2 | 224 | ||
BC048528.1 | - | 2 | 228 | 6774214 |
Gene Symbol: Gm6408 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243104.1 | NP_001230033.1 | 2 | 245 |