Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fastkd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027123.4 | NP_081399.3 | 5 | 1863 | ||
GenBank mRNA | AK009264.1 | - | 5 | 622 | ||
AK031142.1 | - | 5 | 1850 | |||
AK032530.1 | - | 5 | 1946 | |||
AK033189.1 | - | 5 | 1864 | |||
AK036778.1 | - | 5 | 1946 | |||
AK148902.1 | - | 5 | 1854 | |||
BC042506.1 | - | 5 | 1937 | 4023412 | ||
BC060688.1 | - | 4 | 4576 | 6409274 |