Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Exosc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163570.1 | NP_001157042.1 | 6, 7 | 690 | ||
NM_027148.3 | NP_081424.3 | 6, 7 | 678 | |||
GenBank mRNA | AJ505005.1 | - | 6, 7 | 338 | ||
AK009584.1 | - | 6, 7 | 350 | |||
AK169719.1 | - | 4, 5 | 281 | |||
AK220227.1 | - | 6, 7 | 678 | |||
BC059089.1 | - | 6, 7 | 707 | 6509820 | ||
BC067250.1 | - | 5, 6 | 321 |