Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027159.2 | NP_081435.1 | 5 | 1660 | ||
GenBank mRNA | AK010002.1 | - | 5 | 1354 | ||
AK151074.1 | - | 2 | 1867 | |||
AK154179.1 | - | 5 | 1388 | |||
BC019430.1 | - | 5 | 1656 | 3491825 |