Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023524.2 | NP_076013.1 | 5, 6 | 992 | ||
GenBank mRNA | AB035134.1 | - | 5, 6 | 683 | ||
AF267988.1 | - | 5, 6 | 947 | |||
AK010279.1 | - | 5, 6 | 650 | |||
AK177983.1 | - | 2, 3 | 286 | |||
BC024356.1 | - | 5, 6 | 891 | 5066740 |