Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Trip13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027182.2 | NP_081458.1 | 3 | 518 | ||
GenBank mRNA | AK010336.1 | - | 3 | 480 | ||
AK142463.1 | - | 491 | ||||
AK146877.1 | - | 3 | 519 | |||
AK151568.1 | - | 3 | 454 | |||
BC064315.1 | - | 3 | 441 | |||
BC126946.1 | - | 3 | 417 | 40090292 |