Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trip13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027182.2 | NP_081458.1 | 9 | 1008 | ||
GenBank mRNA | AK142463.1 | - | 981 | |||
AK146877.1 | - | 9 | 1009 | |||
AK151568.1 | - | 9 | 944 | |||
AK212754.1 | - | 1 | 46 | |||
AK215913.1 | - | 1 | 68 | |||
BC023834.1 | - | 3 | 216 | |||
BC064315.1 | - | 9 | 931 | |||
BC126946.1 | - | 9 | 907 | 40090292 |