Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027194.3 | NP_081470.2 | 4 | 599 | ||
GenBank mRNA | AK010545.1 | - | 4 | 531 | ||
AK045768.1 | - | 4 | 558 | |||
AK080854.1 | - | 4 | 527 | |||
AK083654.1 | - | 4 | 520 | |||
AK085975.1 | - | 4 | 551 | |||
AK086015.1 | - | 4 | 535 | |||
AK086059.1 | - | 4 | 554 | |||
AK088246.1 | - | 4 | 1064 | |||
AK149407.1 | - | 4 | 528 | |||
AK160039.1 | - | 4 | 536 | |||
AK166012.1 | - | 4 | 552 | |||
AK206565.1 | - | 3 | 178 | |||
BC026789.1 | - | 4 | 526 | 4188168 |