Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh16a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145954.1 | NP_666066.1 | 17 | 2417 | ||
GenBank mRNA | AK010387.1 | - | 4 | 520 | ||
AK030673.1 | - | 17 | 2410 | |||
AK144967.1 | - | 2416 | ||||
AK144995.1 | - | 17 | 2424 | |||
AK151691.1 | - | 17 | 2417 | |||
AK153129.1 | - | 10 | 1324 | |||
AK161198.1 | - | 17 | 2455 | |||
AK169346.1 | - | 2768 | ||||
AK170276.1 | - | 17 | 2456 | |||
AK220200.1 | - | 17 | 2393 | |||
BC013548.1 | - | 17 | 2417 | 4222058 | ||
BC030467.1 | - | 4 | 625 |